missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutarimide, 98%, Thermo Scientific Chemicals

$114.10 - $723.48

Chemical Identifiers

CAS 1121-89-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00006670
InChI Key KNCYXPMJDCCGSJ-UHFFFAOYSA-N
Synonym glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
PubChem CID 70726
ChEBI CHEBI:5435
IUPAC Name piperidine-2,6-dione
SMILES O=C1CCCC(=O)N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0096806
View Documents
Thermo Scientific Chemicals
L0096806
5 g
Each for $114.10
 
AAL0096814
View Documents
Thermo Scientific Chemicals
L0096814
25 g
Each for $288.00
 
AAL0096822
View Documents
Thermo Scientific Chemicals
L0096822
100 g
Each for $723.48
 
Description

Description

Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

1121-89-7
113.12
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
70726
piperidine-2,6-dione
C5H7NO2
MFCD00006670
glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
CHEBI:5435
O=C1CCCC(=O)N1

Specifications

1121-89-7
Nearly Odorless
MFCD00006670
110052
Soluble in water,hot ethanol and boiling benzene. Insoluble in ether.
O=C1CCCC(=O)N1
113.12
CHEBI:5435
98%
153°C to 157°C
C5H7NO2
5 g
glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
piperidine-2,6-dione
70726
113.12
Glutarimide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-340-4

RTECSNumber : MA4000000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only