missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glutaryl dichloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119990250

 View more versions of this product

Catalog No. AC119990250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glutaryl dichloride
2873-74-7
1.3240g/mL
106°C
97%
C5H6Cl2O2
ClCO(CH2)3COCl
25 mL
1.324
Solubility in water: reacts with water.
C(CC(=O)Cl)CC(=O)Cl
169.01
169.01
Liquid
97%
Brown-Red or Colorless to Yellow
216.0°C to 218.0°C
Authentic
Glass bottle
1.4720 to 1.4760
MFCD00000755
02, 634
glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride
YVOFTMXWTWHRBH-UHFFFAOYSA-N
pentanedioyl dichloride
17887
97%

Chemical Identifiers

2873-74-7
169.01
YVOFTMXWTWHRBH-UHFFFAOYSA-N
17887
C(CC(=O)Cl)CC(=O)Cl
C5H6Cl2O2
MFCD00000755
glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride
pentanedioyl dichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 220-711-1

RTECSNumber : SA2583000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only