missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ 4 Fast Flow Media

Single-step purification of GST-tagged proteins and other glutathione S-transferase or glutathione-dependent proteins. Glutathione Sepharose 4 Fast Flow Media is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins.

Supplier:  Cytiva 17513201

 Afficher plus de versions de ce produit

Numéro de catalogue. 45-000-285


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Glutathione coupled to Sepharose 4 Fast Flow via 10-carbon spacer arm
  • Protein binding capacity: ≈10mg recombinant GST/mL media (flowrate dependent)
  • Binding capacity and high flow properties allow scale-up
  • Mild elution conditions preserve protein antigenicity and function
  • Stable in all commonly used aqueous buffers
  • pH stability: 3 to 12

Spécifications

Spécifications

>10 mg/mL
<450 cm/hr.
Highly cross-linked 4% agarose
4% Highly Cross-linked Agarose
base matrix 150-250 cm/h, 100 kPa, XK 50/60 column, bed height 25 cm
25 mL
Glutathione and 10-carbon linker arm
Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed.
10 Carbon Spacer Arm, 4% Agarose, 90 μm Particle Size
3 to 12
4°C to 30°C, 20% Ethanol
90 μm
Chromatography Media
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions