missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol Formal (=1,3-Dioxan-5-ol) 98.0+%, TCI America™

Supplier:  TCI America G0391500MG

Catalog No. G0391500MG


Explore available promotions

Description

Description

Specifications

Specifications

Glycerol Formal (=1,3-Dioxan-5-ol)
Colorless
C4H8O3
500 mg
VCKSNYNNVSOWEE-UHFFFAOYSA-N
1,3-dioxan-5-ol
78475
≥98.0% (GC)
4740-78-7
194°C
MFCD10039829
5-hydroxy-1,3-dioxane, 1,3-formalglycerol, unii-f6up32gbii, glycerol formal, alpha,alpha', f6up32gbii, 5-m-dioxanol, m-dioxan-5-ol, 5-hydroxy-m-dioxane, meta-dioxan-5-ol, ksc498e6n
C1C(COCO1)O
104.105
104.11
Liquid

Chemical Identifiers

4740-78-7
104.105
VCKSNYNNVSOWEE-UHFFFAOYSA-N
78475
C1C(COCO1)O
C4H8O3
MFCD10039829
5-hydroxy-1,3-dioxane, 1,3-formalglycerol, unii-f6up32gbii, glycerol formal, alpha,alpha', f6up32gbii, 5-m-dioxanol, m-dioxan-5-ol, 5-hydroxy-m-dioxane, meta-dioxan-5-ol, ksc498e6n
1,3-dioxan-5-ol
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions