missing translation for 'onlineSavingsMsg'
Learn More

Glycerol formal, 99+%, mixture of isomers, stabilized

CAS: 4740-78-7 | C4H8O3 | 104.11 g/mol

$51.24 - $223.19

Specifications

Chemical Name or Material Glycerol formal
Name Note 99+%
CAS 4740-78-7,5464-28-8
Density 1.2100g/mL
pH 4.0 to 6.5 (10% soln.)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259770050
View Documents
Thermo Scientific Chemicals
259770050
5 mL Glass bottle
Each for $51.24
Only null left
 
AC259772500
View Documents
Thermo Scientific Chemicals
259772500
250 mL Glass bottle
Each for $223.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glycerol formal
4740-78-7,5464-28-8
4.0 to 6.5 (10% soln.)
97°C
99% min. Sum of alpha-and beta-isomers (GC)
C4H8O3
MFCD00003218
1.21
104.11
104.11
Liquid
99+%
1.2100g/mL
193.0°C to 195.0°C
Authentic
Glass bottle
1.4500 to 1.4520
14, 4485
(50% in water) Clear
16 mPa.s (20°C)
99+%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only