Learn More
Glycerol (Molecular Biology), Fisher BioReagents™
For low-temperature storage of enzymes and bacterial cultures
Supplier: Fisher BioReagents BP2294
Description
For low-temperature storage of enzymes and bacterial cultures
Glycerol is used for low-temperature storage of enzymes and bacterial cultures, purification of proteins, and as a component in electrophoresis loading buffers.Specifications
Glycerol | |
>290°C | |
1.261g/cm³ | |
290°C | |
≥99.5 % | |
C3H8O3 | |
MFCD00004722 | |
15, 4520 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
92.09 | |
≥99.5% | |
Viscous Liquid |
56-81-5 | |
18°C | |
5 | |
DNase-, RNase- and Protease-Free | |
HDPE Bottle | |
HOCH2CH(OH)CH2OH | |
4 L | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
OCC(O)CO | |
92.09 | |
CHEBI:17754 | |
0.003mbar at 50°C | |
Molecular Biology |
Chemical Identifiers
56-81-5 | |
92.09 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
OCC(O)CO |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT