missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycerol monostearate, purified, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043883A1

 View more versions of this product

Catalog No. AA43883A1


Explore available promotions

Description

Description

Glycerol monostearate is used as an emulsifier, resin lubricant, opacifier, emollient, bodying agent in a variety of cosmetic formulations for skincare and haircare. It is also used as a thickening, anti-caking and preservative agent. It is also useful for preventing ice creams from drying out or being too sweet. It is further used as a foaming agent for the foam-mat drying of papaya. It is also used as an anti-staling agent in bread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Glycerol monostearate
31566-31-1
MFCD00036186
14,4489
Soluble in hot organic solvents;Soluble in hot water; Slightly soluble in ethanol; Insoluble in aliphatic solvents.
CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O
358.563
CHEBI:75555
Powder
Purified
C21H42O4
1 kg
glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester
VBICKXHEKHSIBG-UHFFFAOYSA-N
2,3-dihydroxypropyl octadecanoate
24699
358.56

Chemical Identifiers

31566-31-1
358.563
VBICKXHEKHSIBG-UHFFFAOYSA-N
24699
2,3-dihydroxypropyl octadecanoate
C21H42O4
MFCD00036186
glyceryl monostearate, monostearin, glyceryl stearate, 1-stearoyl-rac-glycerol, tegin, 1-monostearin, 1-glyceryl stearate, glycerol monostearate, 1-monostearoylglycerol, octadecanoic acid, 2,3-dihydroxypropyl ester
CHEBI:75555
CCCCCCCCCCCCCCCCCC(=O)OCC(CO)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 250-705-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only