Learn More
Glycerol triacetate, 99%, Thermo Scientific Chemicals
$75.62 - $219.14
Chemical Identifiers
CAS | 102-76-1 |
---|---|
Molecular Formula | C9H14O6 |
Molecular Weight (g/mol) | 218.205 |
MDL Number | MFCD00008716 |
InChI Key | URAYPUMNDPQOKB-UHFFFAOYSA-N |
Synonym | triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine |
PubChem CID | 5541 |
ChEBI | CHEBI:9661 |
IUPAC Name | 2,3-diacetyloxypropyl acetate |
SMILES | CC(=O)OCC(COC(=O)C)OC(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1713236
|
Thermo Scientific Chemicals
A1713236 |
500 g |
Each for $75.62
|
|
||||||
AAA171320E
|
Thermo Scientific Chemicals
A171320E |
2500 g |
Each for $219.14
|
|
||||||
Description
Glycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycerol triacetate is used as a food additive and as a solvent in flavorings. It acts as an excipient in pharmaceutical products where it is used as a humectant and a plasticizer. It is also used as a fuel additive, as an antiknock agent, adhesives and sealant chemicals, fillers, intermediates and process regulators.
Solubility
Slightly miscible with water, carbon tetrachloride and carbondisulfide. Miscible with acetone, ethanol, benzene and chloroform.
Notes
Heat sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
102-76-1 | |
218.205 | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
5541 | |
2,3-diacetyloxypropyl acetate |
C9H14O6 | |
MFCD00008716 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
CHEBI:9661 | |
CC(=O)OCC(COC(=O)C)OC(=O)C |
Specifications
102-76-1 | |
1.155 g/mL | |
149°C (300°F) | |
C9H14O6 | |
500 g | |
Slightly miscible with water,carbon tetrachloride and carbondisulfide. Miscible with acetone,ethanol,benzene and chloroform. | |
CC(=O)OCC(COC(=O)C)OC(=O)C | |
218.205 | |
CHEBI:9661 | |
99% |
2°C to 4°C | |
257°C to 259°C | |
99% | |
MFCD00008716 | |
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine | |
URAYPUMNDPQOKB-UHFFFAOYSA-N | |
2,3-diacetyloxypropyl acetate | |
5541 | |
218.21 | |
Glycerol triacetate |
Safety and Handling
EINECSNumber : 203-051-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only