missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
β-Glycerophosphoric acid, disodium salt pentahydrate, 98%, Thermo Scientific Chemicals
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8
$95.49 - $457.63
Chemical Identifiers
CAS | 13408-09-8 |
---|---|
Molecular Formula | C3H19Na2O11P |
Molecular Weight (g/mol) | 308.13 |
MDL Number | MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 |
InChI Key | VWVLGHQKEJESPW-UHFFFAOYSA-N |
Synonym | unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate |
PubChem CID | 131846916 |
IUPAC Name | 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate |
SMILES | O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC410990250
|
Thermo Scientific Chemicals
410990250 |
25 g | Glass bottle |
Each for $95.49
|
|
||||
AC410991000
|
Thermo Scientific Chemicals
410991000 |
100 g | Glass bottle |
Each for $116.14
|
|
||||
AC410995000
|
Thermo Scientific Chemicals
410995000 |
500 g | Glass bottle |
Each for $457.63
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13408-09-8 | |
308.13 | |
VWVLGHQKEJESPW-UHFFFAOYSA-N | |
131846916 | |
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O |
C3H19Na2O11P | |
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate |
Specifications
13408-09-8,13408-09-8 | |
100.0 | |
White | |
98% | |
C3H19Na2O11P | |
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
VWVLGHQKEJESPW-UHFFFAOYSA-N | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate | |
131846916 | |
98% | |
β-Glycerophosphoric acid, disodium salt pentahydrate |
97.5 | |
102.0°C to 104.0°C | |
Authentic | |
Glass bottle | |
CH2OHCH(OPO3Na2)CH2OH·5H2O | |
25 g | |
Solubility in water: freely soluble. | |
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O | |
308.13 | |
306.11 | |
Crystalline Powder |
Safety and Handling
RUO – Research Use Only