missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl Ascorbate 95.0+%, TCI America™
$164.03 - $548.03
Chemical Identifiers
| CAS | 1120360-13-5 |
|---|---|
| Molecular Formula | C9H14O8 |
| Molecular Weight (g/mol) | 250.20 |
| MDL Number | MFCD28386108 |
| InChI Key | KQWQJCDIYBPYNT-UHFFFAOYNA-N |
| Synonym | 2-O-(2,3-Dihydroxypropyl)-L-ascorbic Acid |
| PubChem CID | 133556317 |
| IUPAC Name | 5-(1,2-dihydroxyethyl)-3-(2,3-dihydroxypropoxy)-4-hydroxy-2,5-dihydrofuran-2-one |
| SMILES | OCC(O)COC1=C(O)C(OC1=O)C(O)CO |
Chemical Identifiers
| 1120360-13-5 | |
| 250.20 | |
| KQWQJCDIYBPYNT-UHFFFAOYNA-N | |
| 133556317 | |
| OCC(O)COC1=C(O)C(OC1=O)C(O)CO |
| C9H14O8 | |
| MFCD28386108 | |
| 2-O-(2,3-Dihydroxypropyl)-L-ascorbic Acid | |
| 5-(1,2-dihydroxyethyl)-3-(2,3-dihydroxypropoxy)-4-hydroxy-2,5-dihydrofuran-2-one |
Specifications
| 1120360-13-5 | |
| White-Yellow | |
| MFCD28386108 | |
| 2-O-(2,3-Dihydroxypropyl)-L-ascorbic Acid | |
| OCC(O)COC1=C(O)C(OC1=O)C(O)CO | |
| 250.20 | |
| 250.20 | |
| Crystalline Powder |
| 159°C | |
| C9H14O8 | |
| 5 g | |
| KQWQJCDIYBPYNT-UHFFFAOYNA-N | |
| 5-(1,2-dihydroxyethyl)-3-(2,3-dihydroxypropoxy)-4-hydroxy-2,5-dihydrofuran-2-one | |
| 133556317 | |
| ≥95.0% (T) | |
| Glyceryl Ascorbate |
Safety and Handling
TSCA : No