missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glyceryl Monooleate, Spectrum™ Chemical

G1017, 25496-72-4, C21H40O4

Supplier:  Spectrum Chemical Mfg Cor G1017200LTBL

 View more versions of this product

Catalog No. 18602994

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Spectrum™ Chemical Glyceryl Monooleate is a clear or light yellow oil that is used as an antifoam in juice processing. It has been used as an emulsifier, a moisturizer, and a flavoring agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Specifications

25496-72-4
Drum
MFCD00067258,MFCD00042735,MFCD00069635
RZRNAYUHWVFMIP-KTKRTIGZNA-N
2,3-dihydroxypropyl (9Z)-octadec-9-enoate
Reagent
100%
C21H40O4
200 L
CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO
356.55

Chemical Identifiers

25496-72-4
356.55
RZRNAYUHWVFMIP-KTKRTIGZNA-N
CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO
C21H40O4
MFCD00067258,MFCD00042735,MFCD00069635
2,3-dihydroxypropyl (9Z)-octadec-9-enoate
Videos
SDS
Documents

Documents

Promotions

Promotions