missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine anhydride, 98%, Thermo Scientific Chemicals

$39.50 - $308.41

Chemical Identifiers

CAS 106-57-0
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006009
InChI Key BXRNXXXXHLBUKK-UHFFFAOYSA-N
Synonym 2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
PubChem CID 7817
ChEBI CHEBI:16535
IUPAC Name piperazine-2,5-dione
SMILES C1C(=O)NCC(=O)N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120080050
SDS View Documents
Thermo Scientific Chemicals
120080050
5 g Glass bottle
Each for $39.50
 
AC120080250
SDS View Documents
Thermo Scientific Chemicals
120080250
25 g Glass bottle
Each for $119.15
 
AC120081000
SDS View Documents
Thermo Scientific Chemicals
120081000
100 g Glass bottle
Each for $308.41
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-57-0
114.1
BXRNXXXXHLBUKK-UHFFFAOYSA-N
7817
piperazine-2,5-dione
C4H6N2O2
MFCD00006009
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
CHEBI:16535
C1C(=O)NCC(=O)N1

Specifications

106-57-0
White to Yellow
Authentic
Glass bottle
MFCD00006009
24, 264
2,5-piperazinedione, 2,5-dioxopiperazine, 2,5-diketopiperazine, cyclodiglycine, cycloglycylglycine, cyclo glycylglycyl, glycylglycine lactam, diglycolyl diamide, glycine, bimol. cyclic peptide, alpha,gamma-diacipiperazine
BXRNXXXXHLBUKK-UHFFFAOYSA-N
piperazine-2,5-dione
7817
114.1
Crystalline Powder
>300.0°C
0.5% max. (105°C, 4 hrs)
97.5% min. (ex N)
C4H6N2O2
5 g
15, 7578
Solubility in water: 142g/L water (20 c). Other solubilities: soluble in hcl, slightly soluble in ethanol
C1C(=O)NCC(=O)N1
114.1
CHEBI:16535
98%
Glycine anhydride, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-411-5

RTECSNumber : TL6325250

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only