missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine (Crystalline/Certified), Fisher Chemical™

For amino acid nitrogen standards

Supplier:  Thermo Fisher Scientific G461

 View more versions of this product

Catalog No. G46-1



Explore available promotions

Description

Description

Specifications

Specifications

182°C to 236°C
56-40-6
White
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
75.07
CHEBI:15428
Certified
1 kg
Glycine
5.9 to 6.4
Pass Test
NH2CH2COOH
Poly Bottle
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
750
75.07
1.595g/cm³
Powder

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:1
Instability:1

SDS
Documents

Documents

Promotions

Promotions