missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine ethyl ester hydrochloride, 99%
CAS: 623-33-6 | C4H9NO2·HCl | 139.58 g/mol
$50.73 - $50.73
Chemical Identifiers
| CAS | 623-33-6 |
|---|---|
| Molecular Formula | C4H9NO2·HCl |
| Molecular Weight (g/mol) | 139.58 |
| InChI Key | TXTWXQXDMWILOF-UHFFFAOYSA-N |
| Synonym | glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor |
| PubChem CID | 2723640 |
| IUPAC Name | ethyl 2-aminoacetate;hydrochloride |
| SMILES | CCOC(=O)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC120091000
|
Thermo Scientific Chemicals
120091000 |
100 g | Plastic bottle |
Each for $50.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 623-33-6 | |
| 139.58 | |
| glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor | |
| ethyl 2-aminoacetate;hydrochloride |
| C4H9NO2·HCl | |
| TXTWXQXDMWILOF-UHFFFAOYSA-N | |
| 2723640 | |
| CCOC(=O)CN.Cl |
Specifications
| 623-33-6 | |
| White | |
| 0.5% max. | |
| 99% | |
| C4H9NO2·HCl | |
| 100 g | |
| glycine ethyl ester hydrochloride, ethyl glycinate hydrochloride, ethyl 2-aminoacetate hydrochloride, h-gly-oet.hcl, glycine, ethyl ester, hydrochloride, ethyl aminoacetate hydrochloride, glycine hydrochloride ethyl ester, amino-acetic acid ethyl ester hydrochloride, glycine ethyl ester hydrochloric acid, glycineethylesterhydrochlor | |
| TXTWXQXDMWILOF-UHFFFAOYSA-N | |
| ethyl 2-aminoacetate;hydrochloride | |
| 2723640 | |
| 99% | |
| Glycine ethyl ester hydrochloride |
| 145°C to 148°C | |
| ≥3 (1% aq. soln.) | |
| Authentic | |
| Plastic bottle | |
| H2NCH2CO2C2H5·HCl | |
| 04,34 | |
| Solubility in water: >1000g/L (20°C). Other solubilities: soluble in ethanol | |
| CCOC(=O)CN.Cl | |
| 139.58 | |
| 139.58 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 210-787-4
RUO – Research Use Only