missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine, Multi-Compendial, U.S.P., J.T. Baker™

Supplier:  Avantor J.T.Baker 058201

 View more versions of this product

Catalog No. 02002950


Explore available promotions

Description

Description

Specifications

Specifications

Glycine
C2H5NO2
Wide Mouth Amber Glass Bottle
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
750
75.07
500 g
56-40-6
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
75.07
CHEBI:15428
Multi-Compendial/USP
1L = 1.16kg

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O
Videos
SDS
Documents

Documents

Promotions

Promotions