missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine-N-methylamide hydrochloride, 97%, Thermo Scientific™

$204.58 - $204.58

Chemical Identifiers

CAS 49755-94-4
Molecular Formula C3H9ClN2O
Molecular Weight (g/mol) 124.568
MDL Number MFCD00153440
InChI Key ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
Synonym 2-amino-n-methylacetamide hydrochloride, h-gly-nhme hcl, h-gly-nhme hydrochloride, acetamide, 2-amino-n-methyl-, monohydrochloride, h-gly-nhme.hcl, glycine methyl amide hcl, glycin-methylamid-hydrochlorid, 2-amino-n-methylacetamide-hcl, n-methylglycinamide hydrochloride, glycine methyl amide hydrochloride
PubChem CID 13842202
IUPAC Name 2-amino-N-methylacetamide;hydrochloride
SMILES CNC(=O)CN.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6536706
View Documents
Thermo Scientific Chemicals
H6536706
5 g
Each for $204.58
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

49755-94-4
124.568
ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
13842202
CNC(=O)CN.Cl
C3H9ClN2O
MFCD00153440
2-amino-n-methylacetamide hydrochloride, h-gly-nhme hcl, h-gly-nhme hydrochloride, acetamide, 2-amino-n-methyl-, monohydrochloride, h-gly-nhme.hcl, glycine methyl amide hcl, glycin-methylamid-hydrochlorid, 2-amino-n-methylacetamide-hcl, n-methylglycinamide hydrochloride, glycine methyl amide hydrochloride
2-amino-N-methylacetamide;hydrochloride

Specifications

49755-94-4
C3H9ClN2O
MFCD00153440
2-amino-n-methylacetamide hydrochloride, h-gly-nhme hcl, h-gly-nhme hydrochloride, acetamide, 2-amino-n-methyl-, monohydrochloride, h-gly-nhme.hcl, glycine methyl amide hcl, glycin-methylamid-hydrochlorid, 2-amino-n-methylacetamide-hcl, n-methylglycinamide hydrochloride, glycine methyl amide hydrochloride
CNC(=O)CN.Cl
5 g
13842202
97%
Glycine-N-methylamide hydrochloride
White
NH2CH2CONHCH3·HCl
3678660
ZYZNZBKTJDYJKQ-UHFFFAOYSA-N
2-amino-N-methylacetamide;hydrochloride
124.568
124.57
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'