missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™ Edge
Edge

Supplier:  Fisher BioReagents BP3811

 View more versions of this product

Catalog No. BP381-1



Edge
Explore available promotions

Description

Description

Glycine (White Crystals or Crystalline Powder)

Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.
Specifications

Specifications

182°C
0.1 AU max. at 280nm
56-40-6
5.9 to 6.4
0.003% max.
0.10% max.
7ppm max.
98.5 to 101.0 %
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
1.595g/cm³
2-aminoacetic acid
1 kg
CHEBI:15428
98.5 to 101.0%
Glycine
0.02% max.
1ppm max.
White
10ppm max.
Pass Test
0.20% max.
Pass Test
Poly Bottle
15, 4526
DHMQDGOQFOQNFH-UHFFFAOYSA-N
NCC(O)=O
75.07
750
75.07
Powder

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:1
Instability:1

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions