missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Gray.gif)
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Supplier: Fisher BioReagents BP381500
Description
Glycine (White Crystals or Crystalline Powder)
Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.Specifications
182°C | |
0.1 AU max. at 280nm | |
56-40-6 | |
5.9 to 6.4 | |
0.003% max. | |
98.5 to 101.0 % | |
Pass Test | |
MFCD00008131 | |
0.20% max. | |
Pass Test | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
NCC(O)=O | |
75.07 | |
CHEBI:15428 | |
98.5 to 101.0% | |
500 g |
Glycine | |
0.02% max. | |
1ppm max. | |
White | |
10ppm max. | |
0.10% max. | |
C2H5NO2 | |
7ppm max. | |
15, 4526 | |
Poly Bottle | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
2-aminoacetic acid | |
750 | |
75.07 | |
1.595g/cm³ | |
Powder |
Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Gray.gif)
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : RT