missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glycolaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

$72.78 - $202.73

Chemical Identifiers

CAS 30934-97-5
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00051799
InChI Key NYPNCQTUZYWFGG-UHFFFAOYSA-N
Synonym glycolaldehyde dimethyl acetal, ethanol, 2,2-dimethoxy, unii-u5use33y47, 2,2-dimethoxy-ethanol, 2,2-dimethoxy ethanol, glycolaldehydedimethylacetal, acmc-20aksv, 2,2-dimethoxyethan-1-ol, hydroxyacetaldehyde dimethylacetal, 2-hydroxyacetaldehyde dimethyl acetal
PubChem CID 542381
IUPAC Name 2,2-dimethoxyethanol
SMILES COC(CO)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0709903
View Documents
Thermo Scientific Chemicals
L0709903
1 g
Each for $72.78
 
AAL0709906
View Documents
Thermo Scientific Chemicals
L0709906
5 g
Each for $202.73
 
Description

Description

Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

30934-97-5
106.12
NYPNCQTUZYWFGG-UHFFFAOYSA-N
542381
COC(CO)OC
C4H10O3
MFCD00051799
glycolaldehyde dimethyl acetal, ethanol, 2,2-dimethoxy, unii-u5use33y47, 2,2-dimethoxy-ethanol, 2,2-dimethoxy ethanol, glycolaldehydedimethylacetal, acmc-20aksv, 2,2-dimethoxyethan-1-ol, hydroxyacetaldehyde dimethylacetal, 2-hydroxyacetaldehyde dimethyl acetal
2,2-dimethoxyethanol

Specifications

30934-97-5
1.05
66°C (150°F)
1.413
1 g
glycolaldehyde dimethyl acetal, ethanol, 2,2-dimethoxy, unii-u5use33y47, 2,2-dimethoxy-ethanol, 2,2-dimethoxy ethanol, glycolaldehydedimethylacetal, acmc-20aksv, 2,2-dimethoxyethan-1-ol, hydroxyacetaldehyde dimethylacetal, 2-hydroxyacetaldehyde dimethyl acetal
NYPNCQTUZYWFGG-UHFFFAOYSA-N
2,2-dimethoxyethanol
542381
98%
<-76°C
68°C (21 mmHg)
C4H10O3
MFCD00051799
1697583
Miscible with water.
COC(CO)OC
106.12
106.12
Glycolaldehyde dimethyl acetal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 250-398-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only