missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycolic Acid, Technical, 70-72%, Spectrum™ Chemical
GL185, C2H4O3
$370.74 - $14174.53
Chemical Identifiers
CAS | 79-14-1 |
---|---|
Molecular Formula | C2H4O3 |
Molecular Weight (g/mol) | 76.05 |
InChI Key | AEMRFAOFKBGASW-UHFFFAOYSA-N |
IUPAC Name | 2-hydroxyacetic acid |
SMILES | OCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18603032
|
Spectrum Chemical Mfg Cor
GL185500MLGL |
500 mL | Amber Glass Bottle |
Each for $370.74
|
|
||||
18603033
|
Spectrum Chemical Mfg Cor
GL1854LTGL |
4 L | Amber Glass Bottle |
Each for $1,017.79
|
|
||||
18603031
|
Spectrum Chemical Mfg Cor
GL18520LTPL |
20 L | Poly Drum |
Each for $2,790.47
|
|
||||
18603034
|
Spectrum Chemical Mfg Cor
GL185200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Glycolic Acid, 70 Percent, Technical , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.Chemical Identifiers
79-14-1 | |
76.05 | |
2-hydroxyacetic acid |
C2H4O3 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
OCC(O)=O |
Specifications
79-14-1,7732-18-5 | |
C2H4O3 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
2-hydroxyacetic acid | |
70 to 72% | |
500 mL |
70%,30% | |
UN3265 | |
OCC(O)=O | |
76.05 | |
Technical | |
Amber Glass Bottle |