missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Glycylglycine, 99+%
Supplier: Thermo Scientific Chemicals 120140250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferSpecifications
262.0°C to 264.0°C | |
99+% | |
556-50-3 | |
White | |
Authentic | |
99+% | |
Glass Bottle | |
MFCD00008130 | |
15, 4539 | |
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
2-[(2-aminoacetyl)amino]acetic acid | |
25 g | |
CHEBI:17201 | |
≥99% |
Glycylglycine | |
(10% aq. soln.) (1 cm cell), 0.01 max. at 405nm | |
5.5 to 6 (5% aq. soln.) | |
100ppm max | |
0.5% max. (105°C, 4 hrs) | |
C4H8N2O3 | |
H2NCH2CONHCH2CO2H | |
04, 371 | |
0.1% max. | |
Solubility in water: 132g/L (20°C). Other solubilities: slightly soluble in ethanol, practically insoluble in ether | |
C(C(=O)NCC(=O)O)N | |
132.12 | |
11163 | |
132.12 | |
Fine Crystalline Powder |
Chemical Identifiers
556-50-3 | |
132.12 | |
YMAWOPBAYDPSLA-UHFFFAOYSA-N | |
11163 | |
2-[(2-aminoacetyl)amino]acetic acid |
C4H8N2O3 | |
MFCD00008130 | |
glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid | |
CHEBI:17201 | |
C(C(=O)NCC(=O)O)N |
Safety and Handling
EINECSNumber : 209-127-8
TSCA : TSCA
RUO – Research Use Only