missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Glycylglycylglycylglycine 97.0+%, TCI America™
$109.00 - $667.83
Specifications
Purity | ≥97.0% (HPLC,T) |
---|---|
CAS | 637-84-3 |
Color | White |
Format | Crystal-Powder at 20°C |
Molecular Weight (g/mol) | 246.223 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
G0127100MG
|
TCI America
G0127100MG |
100mg |
Each for $109.00
|
|
|||||
G01271G
|
TCI America
G01271G |
1g |
Each for $667.83
|
|
|||||
Specifications
≥97.0% (HPLC,T) | |
White | |
246.223 |
637-84-3 | |
Crystal-Powder at 20°C | |
Glycylglycylglycylglycine |