Learn More
Glyoxylic acid, 50% in water
CAS: 298-12-4 | C2H2O3 | 74.035 g/mol
$59.46 - $175.50
Chemical Identifiers
| CAS | 298-12-4 |
|---|---|
| Molecular Formula | C2H2O3 |
| Molecular Weight (g/mol) | 74.035 |
| MDL Number | MFCD00006958 |
| InChI Key | HHLFWLYXYJOTON-UHFFFAOYSA-N |
| Synonym | glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid |
| PubChem CID | 760 |
| ChEBI | CHEBI:16891 |
| IUPAC Name | oxaldehydic acid |
| SMILES | C(=O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC411060050
|
Thermo Scientific Chemicals
411060050 |
5 g | Glass bottle |
Each for $59.46
|
|
||||
|
AC411065000
|
Thermo Scientific Chemicals
411065000 |
500 g | Glass bottle |
Each for $175.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 298-12-4 | |
| 74.035 | |
| HHLFWLYXYJOTON-UHFFFAOYSA-N | |
| 760 | |
| oxaldehydic acid |
| C2H2O3 | |
| MFCD00006958 | |
| glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
| CHEBI:16891 | |
| C(=O)C(=O)O |
Specifications
| Colorless to Yellow | |
| 111.0°C | |
| C2H2O3 | |
| MFCD00006958 | |
| 05,320; 07,162; 09,228 | |
| 15, 4546 | |
| Solubility in water: miscible. | |
| C(=O)C(=O)O | |
| 74.035 | |
| CHEBI:16891 | |
| ≥50% | |
| 1.4140 to 1.4180 | |
| 5 g | |
| Glyoxylic acid, 50% in water |
| 298-12-4,7732-18-5 | |
| -93.0°C | |
| HC(O)COOH | |
| 03, IV, 1489 | |
| 1.3000g/mL | |
| glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
| HHLFWLYXYJOTON-UHFFFAOYSA-N | |
| oxaldehydic acid | |
| 760 | |
| 74.04 | |
| Glass bottle | |
| 1.3 | |
| Liquid |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-058-5
RTECSNumber : MD4550000
TSCA : TSCA
RUO – Research Use Only