Learn More
Thermo Scientific Chemicals Glyphosate
$130.71 - $379.83
Chemical Identifiers
CAS | 1071-83-6 |
---|---|
Molecular Formula | C3H8NO5P |
Molecular Weight (g/mol) | 169.07 |
MDL Number | MFCD00055350 |
InChI Key | XDDAORKBJWWYJS-UHFFFAOYSA-N |
IUPAC Name | 2-[(phosphonomethyl)amino]acetic acid |
SMILES | OC(=O)CNCP(O)(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC466180010
|
Thermo Scientific Chemicals
466180010 |
1 g |
Each for $130.71
|
|
|||||
AC466180050
|
Thermo Scientific Chemicals
466180050 |
5 g |
Each for $379.83
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1071-83-6 | |
169.07 | |
XDDAORKBJWWYJS-UHFFFAOYSA-N | |
OC(=O)CNCP(O)(O)=O |
C3H8NO5P | |
MFCD00055350 | |
2-[(phosphonomethyl)amino]acetic acid |
Specifications
1071-83-6 | |
C3H8NO5P | |
Solubility aq. soln.: soluble. Other solubilities: insoluble in most organic solvents | |
XDDAORKBJWWYJS-UHFFFAOYSA-N | |
2-[(phosphonomethyl)amino]acetic acid | |
169.07 |
230.0°C | |
MFCD00055350 | |
1 g | |
OC(=O)CNCP(O)(O)=O | |
169.07 | |
Glyphosate |
Safety and Handling
GHS H Statement Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 213-997-4
RUO – Research Use Only