missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AB222ML
Description
Goat anti-Human IgG Antibody, heavy and light chains is an antibody against Human IgG for use in immunoprecipitation.Specifications
| Goat anti-Human IgG | |
| Polyclonal | |
| Goat serum. Liquid, no preservative. | |
| Human IgG, whole molecule. | |
| Secondary | |
| 2-8°C Lyophilized for up to 12 months from date of receipt −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Protect from light |
| Immunoprecipitation | |
| Unconjugated | |
| Goat | |
| 2 mL | |
| Human | |
| Serum |
For Research Use Only