3M™ Goggle Gear™ 500 Safety Goggles
3M™ Goggle Gear™ Safety Goggles (GG501SGAF-EU) have a clear, polycarbonate lens and 3M™ Scotchgard™ Anti-Fog/Anti-Scratch Coating for superior anti-fog protection.
Supplier:  3M™ 7100079187
Catalog No. 19-169-038
Explore available promotions
Description
Description
  • Modern slim-line and lightweight design for an excellent fit
  • Easy-to-adjust elastic nylon headband for stability and comfort
  • Pivoting ratchet for convenient personal adjustment
  • 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties
  • Coating withstands washing, soaking in bleach or using alcohol wipes without losing performance
  • Special 3M™ Goggle Gear prescription insert available
  • Offers excellent protection against UV radiation
  • Lens Marking: 2C-1.2 3M 1 BT KN
  • Frame Marking: 3M EN 166 3 4 BT CE
  • Protection against large dust particles (3)
  • Protection against liquid droplets (4)
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
Anti-fog
Clear
Red
Vented
ANSI Z87.1-2015, CSA Z94.3
Safety Goggles
GG501SGAF
Polycarbonate