missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03974103

 View more versions of this product

Catalog No. AA3974103


Explore available promotions

Description

Description

Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold(I) sodium thiosulfate hydrate
White
Powder
99.9% (Metal basis)
MFCD00046176
Soluble in water. Insoluble in ethanol and most other organic solvents.
[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
490.17
490.19 (anhydrous)
3.09
(decomposition)
15283-45-1
1 g
AuNa3O6S4
sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
FJCMCHNDCIYVST-UHFFFAOYSA-J
gold(1+) trisodium disulfanidesulfonate
27176
White powder

Chemical Identifiers

15283-45-1
490.17
FJCMCHNDCIYVST-UHFFFAOYSA-J
27176
[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
AuNa3O6S4
MFCD00046176
sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
gold(1+) trisodium disulfanidesulfonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-324-4

RTECSNumber : CK6235000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only