Corning™ Sterile Polyethylene Blender Bags
Universal bags for homogeneous suspension
Supplier:  Corning™ SM101
 View more versions of this product
Catalog No. 07-201-919
Explore available promotions
Description
Description
  • Polyethylene (PE) bags
  • 80 to 3500 mL
  • With or without filter
  • Traceability on each filter bag
  • Sterile
Order Info
Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.
Specifications
Specifications
Without filter
Natural
Sterile
1500/Pk.
150 mm
80 mL
Polyethylene
4500/Cs.
25/Pk.
105 mm