Promega GoTaq™ G2 DNA Polymerase
GoTaq™ G2 DNA Polymerase is supplied with 5X Green GoTaq™ Reaction Buffer and 5X Colorless GoTaq™ Reaction Buffer.
Supplier: Promega M7848
Description
- Next-Generation GoTaq(R) for Robust, Reliable PCR Amplification
- Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
- Direct-to-gel convenience
- 5X Reaction Buffer with MgCl2 to simplify reaction setup
- Supplied at a concentration of 5u/μL
Specifications
−30° to −10°C | |
2500U | |
5U/μL |
DNA Polymerase | |
RT-PCR |