Promega GoTaq™ G2 Flexi DNA Polymerase DFS Item
GoTaq™ G2 Flexi DNA Polymerase is supplied with 5X Green GoTaq™ Flexi Buffer and 5X Colorless GoTaq™ Flexi Buffer and 25mM MgCl2.
Supplier:  Promega M7805
 View more versions of this product
Catalog No. PRM7805
Explore available promotions
Description
Description
  • High-Performance GoTaq(R) G2 DNA Polymerase with Mg-Free Buffer System
  • Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
  • Direct to gel convenience
  • Flexibility to optimize Mg concentration in PCR
  • Supplied at a concentration of 5u/μL
Specifications
Specifications
−30° to −10°C
500U
5U/μL
Flexi DNA Polymerase
RT-PCR