Promega GoTaq™ G2 Flexi DNA Polymerase
GoTaq™ G2 Flexi DNA Polymerase is supplied with 5X Green GoTaq™ Flexi Buffer and 5X Colorless GoTaq™ Flexi Buffer and 25mM MgCl2.
Supplier: Promega M7808
Description
- High-Performance GoTaq(R) G2 DNA Polymerase with Mg-Free Buffer System
- Full-length Taq DNA polymerase that exhibits 5′→3′ exonuclease activity
- Direct to gel convenience
- Flexibility to optimize Mg concentration in PCR
- Supplied at a concentration of 5u/μL
Specifications
−30° to −10°C | |
10.000U | |
5U/μL |
Flexi DNA Polymerase | |
RT-PCR |