missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GoTaq™ G2 Green Master Mix
GoTaq(R) G2 Master Mixes are premixed ready-to-use solutions containing GoTaq(R) G2 DNA Polymerase, dNTPs, MgCl2 and reaction buffers at optimal concentrations for efficient amplification of a wide range of DNA templates by PCR.
$50.51 - $430.41
Specifications
Content And Storage | −30° to −10°C |
---|---|
Description | Green Master Mix |
For Use With (Application) | RT-PCR |
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Includes | Price | Quantity | ||||
PRM7823
|
Promega
M7823 |
1000 reactions | Green Master Mix (M782B 1 x 25mL), Nuclease-Free Water (P119C 1 x 25mL) |
Each for $430.41
|
|
||||
PRM7822
|
Promega
M7822 |
100 reactions | Green Master Mix (M782A 2 x 1.25mL), Nuclease-Free Water (P119A 2 x 1,250μL) |
Each for $50.51
|
|
||||
Description
- All-in-One PCR Master Mix: Just Add Template and Primers
- High-Performance GoTaq(R) G2 DNA Polymerase in a Ready-to-Use Master Mix
- Simplifies reaction setup and saves time
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
Specifications
−30° to −10°C | |
Green Master Mix | |
RT-PCR |