Learn More
Eisco™ Graduated Cylinder, Borosilicate, Class B, Hexagonal Base with Spout, Blue Graduation
The sturdy borosilicate glass graduated cylinder is ideal for measuring liquids. The graduation markings make it easy to read liquid levels and the spout allows for liquids to be poured from the vessel with little spillage.
Supplier:  Eisco™ CH0345E
 View more versions of this product
Catalog No. S23907
Only null left
Explore available promotions
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Hexagonal base for maximum stability
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class B
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Specifications
Specifications
Round
±5 mL
B
Blue
500 mL
Autoclavable
DIN EN ISO 4788
Hexagonal
Graduated Measuring Cylinder
Borosilicate Glass
5 mL
Product Suggestions
Product Suggestions