Eisco™ Graduated Cylinder, Borosilicate, Class B, Hexagonal Base with Spout, Blue Graduation
The sturdy borosilicate glass graduated cylinder is ideal for measuring liquids. The graduation markings make it easy to read liquid levels and the spout allows for liquids to be poured from the vessel with little spillage.
Supplier:  Eisco™ CH0345F
 View more versions of this product
Catalog No. S23908
Explore available promotions
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Hexagonal base for maximum stability
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class B
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Specifications
Specifications
Round
Hexagonal
Graduated Measuring Cylinder
Borosilicate Glass
10 mL
±10 mL
B
Blue
1000 mL
Autoclavable