Graduated Cylinders
With embossed calibrations for easy measuring.
Supplier:  Customer Channels Group (Ccg) 78105
 View more versions of this product
Catalog No. S95594
Explore available promotions
Description
Description
  • Durable, transparent polypropylene
  • Wide, stable bases
  • Chemical and acid resistant
  • Autoclavable (dry heat up to 120°C [248°F], wet heat up to 80°C [176°F])
  • Microwavable
Order Info
Available individually or as seven-piece set.
Specifications
Specifications
Autoclavable
120°C
Wide
Graduated Cylinder