Graduated Cylinders
With embossed calibrations for easy measuring.
Supplier: Customer Channels Group (Ccg) 78115
Description
- Durable, transparent polypropylene
- Wide, stable bases
- Chemical and acid resistant
- Autoclavable (dry heat up to 120°C [248°F], wet heat up to 80°C [176°F])
- Microwavable
Order Info
Available individually or as seven-piece set.
Specifications
Autoclavable | |
120°C | |
Wide | |
Graduated Cylinder |