missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Globe Scientific Graduated Transfer Pipet

Graduated transfer pipets are designed for measurements and feature molded-in graduation marks that will never fade or wash off.

Supplier:  Globe Scientific 137018H

Catalog No. 145634789


Explore available promotions

Description

Description

  • Molded-in graduation marks to 1 mL that will never fade or wash off
Specifications

Specifications

No
No
Sterile
Molded
100 Pk.; 5 Pk./Unit
Clear
Transfer Pipet
Not Autoclavable
LDPE
Yes
150 mm
Individually Wrapped
5 mL
Single-use
SDS
Documents

Documents

Promotions

Promotions