missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Gram Crystal Violet

Can be used as a replacement reagent for the BD™ Gram Stain Kits. BD™ Gram Crystal Violet is used as the primary stain in a 4-step Gram staining process. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.

$277.53 - $446.24

Specifications

Type Crystal Violet
Certifications/Compliance CE
Format Liquid
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Color Purple
View More Specs

Products 2
Catalog Number Mfr. No. Type Quantity Price Quantity  
Catalog Number Mfr. No. Type Quantity Price Quantity  
B4312525
View Documents
BD
212525
Crystal Violet 4 x 250mL
Case of 4 Each for $277.53
 
B4312526
View Documents
BD
212526
Crystal Violet 1 Gallon (3.8L)
Each for $446.24
 
Description

Description

Gram Stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.

  • Complete stain kits reduce the mess associated with Gram staining procedures and include a compact tray for holding the 250mL reagent bottles
  • Break-resistant bottles features no-drip, keep-clean cap spouts that make regulating the flow easy and that help to prevent spots and spills
  • Color-coded labels allow quick identification of the reagent

Order Info

Available in packs of four 250mL bottles or individual one-gallon bottles

Specifications

Specifications

Crystal Violet
Liquid
Purple
Crystal Violet
CE
Used to stain microorganisms from cultures or specimens by the differential gram method
15° to 30°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'