missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Graphite powder, natural, briquetting grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040795TD

 View more versions of this product

Catalog No. AA40795TD


Explore available promotions

Description

Description

Graphite Powder is basically used for casting and polishing purposes, moulding & Carbon addition in steel industries. It is also used for smooth casting iron, copper base alloys and aluminum base alloy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Graphite powder
7782-42-5
−200 Mesh
Odorless
C
454 g
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
99.9995%
Powder
Natural
Black
UCP-1 grade, ultra 'F' purity
(metals basis)
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
UN1325
Insoluble in water.
[C]
12.01
CHEBI:27594
Briquetting

Chemical Identifiers

7782-42-5
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H228-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 231-955-3

RTECSNumber : MD9659600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only