missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Graphite powder, natural, universal grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

$94.42 - $1,082.92

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40799TA
View Documents
Thermo Scientific Chemicals
040799TA
28 g N/A
 
AA40799TB
View Documents
Thermo Scientific Chemicals
040799TB
113 g N/A
 
AA40799TC
View Documents
Thermo Scientific Chemicals
040799TC
227 g N/A
 
AA40799TD
View Documents
Thermo Scientific Chemicals
040799TD
454 g N/A
 
Description

Description

Used in aircraft manufacturing industries, defense material, automotive industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-42-5
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7782-42-5
−200 Mesh
C
28 g
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
99.9995%
Powder
(metals basis)
Black
Odorless
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
UN1325
Insoluble.
[C]
12.01
CHEBI:27594
UCP-1-U grade, ultra 'F' purity
Graphite powder, Natural
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H228-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 231-955-3

RTECSNumber : MD9659600

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only