Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel
For determinations in clinical chemistry and immunochemistry. Greiner Bio-One Plasma Tubes with Lithium Heparin and Gel includes a safety pull cap.
Supplier: Greiner Bio-One 456287P
Description
Plasma stability in a spun tube.
Note: This 3.5mL tube (22-040-534) is a larger sized tube (13 x 100mm), and may not fit certain instruments. For another 3.5mL tube option which is a smaller size (13 x 75mm), see Cat No. 22-040-539.- Interior tube wall is coated with lithium heparin, an anticoagulant that binds antithrombins to block the coagulation cascade producing a plasma sample
- Contain a barrier gel in the tube that moves upward to form a stable barrier separating plasma from cells during centrifugation; plasma may be aspirated directly from the tube eliminating the need for transfer
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
Lithium Heparin | |
3.5 mL | |
13 x 100 mm |
Safety Pull Cap | |
Green with Yellow Ring |