Greiner Bio-One MiniCollect™ Capillary Blood Collection System Accessories
For use with MiniCollect Capillary Blood Collection System
$38.03 - $41.21
Specifications
For Use With (Equipment) | Greiner Vacuette MiniCollect Capillary Blood Collection System |
---|
Catalog Number | Mfr. No. | Product Type | Tube Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Tube Size | Price | Quantity | ||||
22-040-092
|
Greiner Bio-One
450436 |
Lithium Heparin Tube | 80 μL |
Pack of 100 Each for $38.03
Case of 10 Pack for $380.30
|
|
||||
22-040-082
|
Greiner Bio-One
450420 |
Capillary Tube | 13 x 75 mm |
Pack of 100 Each for $39.55
Case of 10 Pack for $395.50
|
|
||||
22-040-081
|
Greiner Bio-One
450419 |
Capillary Tube | 13 x 75 mm |
Pack of 100 Each for $39.55
Case of 10 Pack for $395.50
|
|
||||
22-040-090
|
Greiner Bio-One
450431 |
Serum Tube | 80 μL |
Pack of 100 Each for $41.21
Case of 10 Pack for $412.10
|
|
||||
22-040-083
|
Greiner Bio-One
450421 |
Funnel | — |
Pack of 100 Each for $38.69
Case of 10 Pack for $386.90
|
|
||||
Description
- Capillary or funnel can be inserted into MiniCollect tubes without removing the cap
- 13 × 75mm carrier tubes facilitate bar code labeling, centrifugation, and storage
- Use amber carrier tube for sheilding light-sensitive analytes
Specifications
Greiner Vacuette MiniCollect Capillary Blood Collection System |