Fisherbrand™ Grey Phenolic Work Surface
Fisherbrand™ Phenolic Work Surface is a chemically resistant, one-inch thick grey Phenolic top.
 View more versions of this product
Catalog No. 06000380
Explore available promotions
Description
Description
  • Front edge finished, all other edges are square
  • Curb must be ordered separately
Specifications
Specifications
60 in.
30 in.
76.2 cm
1 in.
Phenolic
Work Surface
2.5 cm
152.4 cm
Gray