missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GSC Go Science Crazy Cast Iron Support Ring Stand
Great for holding all sorts of lab equipment in a small space. The GSC Cast Iron Support Ring Stand is made from cast-iron, with an enamel finish that is acid resistant. Includes a zinc plated steel rod that screws into the base support.
$32.05 - $67.82
Specifications
Type | Stand |
---|---|
Material | Cast Iron |
Alert:
Not a toy. Product intended for ages 13 and up.
Includes: Zinc plated steel rod that screws into the base support
Catalog Number | Mfr. No. | Width (English) | Length (English) | Diameter (English) | Height (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Length (English) | Diameter (English) | Height (English) | Price | Quantity | ||
S13745
|
GSC Go Science Crazy
4SSC14 |
4 in. | 6 in. | 5/16 in. | 18 in. |
Each for $32.05
|
|
||
S13746
|
GSC Go Science Crazy
4SSC15 |
5 in. | 8 in. | 3/8 in. | 20 in. |
Each for $35.31
|
|
||
S13747
|
GSC Go Science Crazy
4SSC16 |
6 in. | 9 in. | 1/2 in. | 24 in. |
Each for $53.56
|
|
||
S13748
|
GSC Go Science Crazy
4SSC17 |
6 in. | 11 in. | 1.2 in. | 36 in. |
Each for $67.82
|
|
||
Description
- Stand made from cast iron
- Acid resistant
- Plated steel support rod
- Rod threaded to fit base
Specifications
Stand | |
Cast Iron |