GSC Go Science Crazy Clamp Holder
A cast iron clamp which can rotate, and is used to hold objects on a support rod
$11.74 - $32.67
Specifications
Type Clamp Holder
Material Cast Iron, Nickel Plated (Screws)
For Use With (Application) For use with GSC Support Stands and Rods
View More Specs
Alert:
Not a toy. Product intended for ages 13 and up.
Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
S13737
View Documents
GSC Go Science Crazy
4ACH40
Adjustable, can be rotated 360 deg. and locked in place
Each for $32.67
Pack of 10 Each for $326.70
S13738
View Documents
GSC Go Science Crazy
4CH39
Accept rods up to 0.625 in. dia.
Each for $11.74
Pack of 10 Each for $117.40
Description
Description
  • To hold objects on support rod
  • Cast iron construction
  • Clamp can rotate 360 degrees and locked in place
  • Nickel-plated thumb screws
  • Accepts rods up to 0.625 in. dia.
Specifications
Specifications
Clamp Holder
Cast Iron, Nickel Plated (Screws)
For use with GSC Support Stands and Rods
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'