GSC Go Science Crazy Gas Measuring Tubes
Allows students to learn about measuring gas.
Supplier:  GSC Go Science Crazy GIG7012
 View more versions of this product
Catalog No. S32366
Explore available promotions
Description
Description
  • Borosilicate glass
  • Permenant graduations
  • 50mL subdivions at 0.1mL and 100mL subdivided at 0.2mL
  • Available in the following sizes:
    • #701-1: 50mL with 0.1 mL subdivisions
    • #701-2: 100mL with 0.2 mL subdivisions
Specifications
Specifications
100 mL
Borosilicate Glass
For measuring gas