GSC Go Science Crazy Support Ring Clamp
Great for holding all sorts of lab equipment in a small space. Works best with GSC Support Stands and Rods. The GSC Support Ring Clamp is a cast-iron clamp with nickel plated steel set screws. Can be clamped to a support stand rod for holding lab equipment.
Supplier:  GSC Go Science Crazy 4SRC48
 View more versions of this product
Catalog No. S13756
Explore available promotions
Alert:
Not a toy. Product intended for ages 13 and up.
Description
Description
  • Clamp made from cast iron
  • Nickel-plated steel screws
  • Great for support stands/rods
Specifications
Specifications
5 in. (Outer Dia.)