missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Guaiacol | |
White | |
C7H8O2 | |
500 g | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
2-methoxyphenol | |
460 | |
124.14 | |
Crystals |
90-05-1 | |
205°C | |
MFCD00002185 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
COC1=CC=CC=C1O | |
124.139 | |
CHEBI:28591 | |
≥98.0% (GC) |
Chemical Identifiers
90-05-1 | |
124.139 | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
460 | |
2-methoxyphenol |
C7H8O2 | |
MFCD00002185 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
CHEBI:28591 | |
COC1=CC=CC=C1O |
Safety and Handling
EINECSNumber : (3)-0567
RTECSNumber : SL7525000
TSCA : Yes