missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guaiacylglycerol-beta-guaiacyl Ether 97.0+%, TCI America™
$346.71 - $2143.67
Chemical Identifiers
| CAS | 7382-59-4 |
|---|---|
| Molecular Formula | C17H20O6 |
| Molecular Weight (g/mol) | 320.34 |
| MDL Number | MFCD00060144 |
| InChI Key | COXIJIHARCXZBW-UHFFFAOYNA-N |
| Synonym | 1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)-1,3-propanediol |
| PubChem CID | 6424189 |
| ChEBI | CHEBI:53650 |
| IUPAC Name | 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol |
| SMILES | COC1=CC(=CC=C1O)C(O)C(OCO)C1=CC=CC=C1OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0233100MG
|
TCI America
G0233100MG |
100 mg |
Each for $346.71
|
|
|||||
|
G02331G
|
TCI America
G02331G |
1 g |
Each for $2,143.67
|
|
|||||
Chemical Identifiers
| 7382-59-4 | |
| 320.34 | |
| COXIJIHARCXZBW-UHFFFAOYNA-N | |
| 6424189 | |
| 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol |
| C17H20O6 | |
| MFCD00060144 | |
| 1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)-1,3-propanediol | |
| CHEBI:53650 | |
| COC1=CC(=CC=C1O)C(O)C(OCO)C1=CC=CC=C1OC |
Specifications
| 7382-59-4 | |
| White | |
| MFCD00060144 | |
| 1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)-1,3-propanediol | |
| COC1=CC(=CC=C1O)C(O)C(OCO)C1=CC=CC=C1OC | |
| 320.34 | |
| CHEBI:53650 | |
| ≥97.0% (GC) | |
| Guaiacylglycerol-beta-guaiacyl Ether |
| 100°C | |
| C17H20O6 | |
| 100 mg | |
| COXIJIHARCXZBW-UHFFFAOYNA-N | |
| 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol | |
| 6424189 | |
| 320.34 | |
| Crystalline Powder |
Safety and Handling
TSCA : No