missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Guaiazulene, 98+%, Thermo Scientific Chemicals
$70.62 - $227.47
Chemical Identifiers
CAS | 489-84-9 |
---|---|
Molecular Formula | C15H18 |
Molecular Weight (g/mol) | 198.309 |
MDL Number | MFCD00003811 |
InChI Key | FWKQNCXZGNBPFD-UHFFFAOYSA-N |
Synonym | guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen |
PubChem CID | 3515 |
ChEBI | CHEBI:5550 |
IUPAC Name | 1,4-dimethyl-7-propan-2-ylazulene |
SMILES | CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1769006
|
Thermo Scientific Chemicals
A1769006 |
5 g |
Each for $70.62
|
|
|||||
AAA1769014
|
Thermo Scientific Chemicals
A1769014 |
25 g |
Each for $227.47
|
|
|||||
Description
It is employed as antimicrobial agent and perfuming agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as antimicrobial agent and perfuming agent in cosmetics industry.
Solubility
Soluble in alcohol, water, 0.1115 mg/L @ 25°C (est).
Notes
Store in dark. Protect from heat, oxidizing agents and heat. Keep it refrigerated.
Chemical Identifiers
489-84-9 | |
198.309 | |
FWKQNCXZGNBPFD-UHFFFAOYSA-N | |
3515 | |
1,4-dimethyl-7-propan-2-ylazulene |
C15H18 | |
MFCD00003811 | |
guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen | |
CHEBI:5550 | |
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C |
Specifications
489-84-9 | |
0.976 | |
>110°C (230°F) | |
MFCD00003811 | |
1365001 | |
14,4554 | |
Soluble in alcohol,water,0.1115mg/L @ 25°C (est). | |
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C | |
198.309 | |
CHEBI:5550 | |
≥98% |
31°C to 33°C | |
152°C to 154°C (7 mmHg) | |
C15H18 | |
5 g | |
Light sensitive | |
guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen | |
FWKQNCXZGNBPFD-UHFFFAOYSA-N | |
1,4-dimethyl-7-propan-2-ylazulene | |
3515 | |
198.31 | |
Guaiazulene |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 207-701-2
RTECSNumber : CO4790000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only